Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2 | NM_001270467.1 | 3 | 1926 | NP_001257396.1 |
| NM_001270468.1 | 3 | 1895 | NP_001257397.1 | |
| NM_001270469.1 | 3 | 1893 | NP_001257398.1 | |
| NM_001270470.1 | 3 | 2165 | NP_001257399.1 | |
| NM_001270471.1 | 2 | 1818 | NP_001257400.1 | |
| NM_003877.4 | 3 | 2036 | NP_003868.1 | |
| XM_011538929.1 | XP_011537231.1 | |||
| XM_011538935.1 | XP_011537237.1 | |||
| XM_011538936.1 | XP_011537238.1 | |||
| XM_017020147.1 | 4 | 1673 | XP_016875636.1 | |
| XM_017020148.1 | 4 | 1563 | XP_016875637.1 | |
| XM_017020149.1 | 4 | 1557 | XP_016875638.1 | |
| XM_017020150.1 | 4 | 1764 | XP_016875639.1 | |
| XM_017020151.1 | 4 | 1569 | XP_016875640.1 | |
| XM_017020152.1 | 4 | 1598 | XP_016875641.1 | |
| XM_017020153.1 | 4 | 1340 | XP_016875642.1 | |
| XM_017020154.1 | 4 | 1402 | XP_016875643.1 | |
| XM_017020155.1 | XP_016875644.1 | |||
| XM_017020156.1 | 3 | 2151 | XP_016875645.1 | |
| AF037989.1 | 3 | 1763 | AAC34745.1 | |
| BC010399.1 | 2 | 1658 | AAH10399.1 | |
| CD699665.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs200120992] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map