Assay Details
Target Gene Details
Entrez Gene ID: | 4750 |
Gene Name: | NIMA related kinase 1 |
Gene Aliases: |
NY-REN-55, SRPS2, SRPS2A, SRTD6 |
Location: |
Chr.4:169393270-169612629 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK1 | NM_001199397.1 | 36 | 4778 | NP_001186326.1 |
| NM_001199398.1 | 35 | 4646 | NP_001186327.1 | |
| NM_001199399.1 | 33 | 4487 | NP_001186328.1 | |
| NM_001199400.1 | 33 | 4640 | NP_001186329.1 | |
| NM_012224.2 | 34 | 4772 | NP_036356.1 | |
| XM_006714228.1 | 35 | 4603 | XP_006714291.1 | |
| XM_011532003.1 | 35 | 4696 | XP_011530305.1 | |
| XM_011532004.1 | 34 | 4564 | XP_011530306.1 | |
| XM_017008249.1 | 35 | 4682 | XP_016863738.1 | |
| XM_017008250.1 | 34 | 4800 | XP_016863739.1 | |
| XM_017008251.1 | 34 | 4598 | XP_016863740.1 | |
| XM_017008252.1 | 32 | 4618 | XP_016863741.1 | |
| XM_017008253.1 | 35 | 4628 | XP_016863742.1 | |
| XM_017008254.1 | 28 | 3738 | XP_016863743.1 | |
| AB067488.1 | 34 | 4608 | ||
| AF155113.1 | 18 | 2656 | AAD42879.1 | |
| AK025658.1 | 9 | 1526 | BAB15207.1 | |
| AL050385.1 | 35 | 4602 | CAI46225.1 | |
| BC037790.2 | 8 | 1438 | AAH37790.1 | |
| BC114491.1 | ||||
| CD358162.1 | 4 | 777 | ||
| CR933642.1 | 36 | 4736 | CAI45943.1 | |
| CR936672.1 | 33 | 4598 |
More Information
Set Membership: |
|

Genomic Map