Assay Details
Target Gene Details
Entrez Gene ID: | 51399 |
Gene Name: | trafficking protein particle complex 4 |
Gene Aliases: |
CGI-104, HSPC172, PTD009, SBDN, SYNBINDIN, TRS23 |
Location: |
Chr.11:119018469-119023674 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC4 | NM_001318486.1 | 4 | 972 | NP_001305415.1 |
| NM_001318488.1 | 5 | 962 | NP_001305417.1 | |
| NM_001318489.1 | 5 | 914 | NP_001305418.1 | |
| NM_001318490.1 | 5 | 947 | NP_001305419.1 | |
| NM_001318492.1 | 5 | 1183 | NP_001305421.1 | |
| NM_001318494.1 | 3 | 797 | NP_001305423.1 | |
| NM_016146.5 | 5 | 1076 | NP_057230.1 | |
| AF078862.1 | 5 | 1006 | AAD44494.1 | |
| AF105025.1 | 5 | 1014 | AAF21897.1 | |
| AF151862.1 | 5 | 1012 | AAD34099.1 | |
| AF161520.1 | 5 | 799 | AAF29135.1 | |
| AK025623.1 | ||||
| AK290520.1 | 5 | 779 | ||
| AK293977.1 | 5 | 665 | ||
| AK297422.1 | 5 | 617 | ||
| BC010866.1 | 5 | 775 | AAH10866.1 | |
| BU624012.1 | 1 | 203 |
More Information
Set Membership: |
|

Genomic Map