Assay Details
Target Gene Details
Entrez Gene ID: | 151647 |
Gene Name: | family with sequence similarity 19 member A4, C-C motif chemokine like |
Gene Aliases: |
TAFA-4, TAFA4 |
Location: |
Chr.3:68731764-68932610 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A4 | NM_001005527.2 | 6 | 1456 | NP_001005527.1 |
| NM_182522.4 | 6 | 1538 | NP_872328.1 | |
| XM_011533371.1 | 6 | 1497 | XP_011531673.1 | |
| XM_011533372.1 | 6 | 1373 | XP_011531674.1 | |
| AK057890.1 | 5 | 1154 | BAB71606.1 | |
| AY325117.1 | 6 | 1510 | AAP92409.1 | |
| BC031566.1 | 6 | 1370 | AAH31566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160944 | Chr.3:68481567 - 68862129 on Build GRCh38 | Duplication |
|
| nsv834722 | Chr.3:68725885 - 68924969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map