Assay Details
Target Gene Details
Entrez Gene ID: | 2896 |
Gene Name: | granulin |
Gene Aliases: |
CLN11, GEP, GP88, PCDGF, PEPI, PGRN |
Location: |
Chr.17:44345086-44353106 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRN | NM_002087.3 | 1 | 80 | NP_002078.1 |
| BC000324.2 | 1 | 43 | AAH00324.1 | |
| DC336362.1 | 1 | 80 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map