Assay Details
Target Gene Details
Entrez Gene ID: | 22950 |
Gene Name: | solute carrier family 4 member 1 adaptor protein |
Gene Aliases: |
HLC3 |
Location: |
Chr.2:27663471-27694980 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A1AP | NM_018158.2 | 6 | 1702 | NP_060628.2 |
| AK001486.1 | 6 | 1702 | BAA91718.1 | |
| AK300958.1 | 8 | 1212 | ||
| AY028435.1 | 6 | 1678 | AAK29177.1 | |
| AY117688.1 | 6 | 1091 | AAN12269.1 | |
| BC098302.1 | 6 | 1697 | AAH98302.1 | |
| BC098358.1 | 6 | 1697 | AAH98358.1 | |
| BC099711.1 | 6 | 1697 | AAH99711.1 | |
| BC099739.1 | 6 | 1695 | AAH99739.1 |
More Information
Set Membership: |
|

Genomic Map