Assay Details
Target Gene Details
Entrez Gene ID: | 84919 |
Gene Name: | protein phosphatase 1 regulatory subunit 15B |
Gene Aliases: |
CREP, MSSGM2 |
Location: |
Chr.1:204398684-204411880 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R15B | NM_032833.4 | 1 | 2109 | NP_116222.4 |
| XM_005245551.4 | 1 | 2172 | XP_005245608.2 | |
| AK027650.1 | 1 | 2109 | BAB55266.1 | |
| AK222877.1 | 1 | 2110 | BAD96597.1 | |
| AK303535.1 | 2 | 1663 | ||
| AL833746.1 | 1 | 786 | CAH56240.1 | |
| BC065280.1 | 1 | 2083 | AAH65280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952641 | Chr.1:204396873 - 204410972 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map