Assay Details
Target Gene Details
Entrez Gene ID: | 4200 |
Gene Name: | malic enzyme 2 |
Gene Aliases: |
ODS1 |
Location: |
Chr.18:50878604-50949795 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ME2 | NM_001168335.1 | NP_001161807.1 | ||
| NM_002396.4 | NP_002387.1 | |||
| AK313391.1 | ||||
| AU119085.1 | ||||
| BC000147.1 | AAH00147.1 | |||
| BI465083.1 | ||||
| BI559761.1 | ||||
| DA644731.1 | ||||
| M55905.1 | AAA36197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893102 | Chr.18:50871431 - 50953868 on Build GRCh38 | Loss |
|
| nsv9630 | Chr.18:50532107 - 50922466 on Build GRCh38 | Loss |
|
| esv3642520 | Chr.18:50904416 - 50913598 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map