Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYB | NM_006166.3 | 5 | 563 | NP_006157.1 |
| XM_005268908.4 | 6 | 627 | XP_005268965.1 | |
| XM_005268909.4 | 6 | 610 | XP_005268966.1 | |
| XM_005268910.4 | 6 | 1282 | XP_005268967.1 | |
| AK291934.1 | 5 | 495 | ||
| BC005316.1 | 5 | 512 | AAH05316.1 | |
| BC005317.1 | 5 | 518 | AAH05317.1 | |
| BC007035.1 | 5 | 490 | AAH07035.1 | |
| BG776259.1 | 5 | 564 | ||
| L06145.1 | 3 | 362 | AAA59930.1 | |
| X59710.1 | 5 | 467 | CAA42230.1 |
More Information
Set Membership: |
|

Genomic Map