Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOPEY1 | XM_011535619.2 | XP_011533921.1 | ||
| XM_017010559.1 | XP_016866048.1 | |||
| XM_017010560.1 | XP_016866049.1 | |||
| XM_017010561.1 | XP_016866050.1 | |||
| XM_017010562.1 | XP_016866051.1 | |||
| XM_017010563.1 | 39 | 8368 | XP_016866052.1 | |
| XM_017010564.1 | XP_016866053.1 | |||
| XM_017010565.1 | 39 | 8362 | XP_016866054.1 | |
| XM_017010566.1 | XP_016866055.1 | |||
| XM_017010567.1 | 38 | 8290 | XP_016866056.1 | |
| XM_017010568.1 | 38 | 8359 | XP_016866057.1 | |
| XM_017010569.1 | 37 | 8276 | XP_016866058.1 | |
| XM_017010570.1 | XP_016866059.1 | |||
| XM_017010571.1 | XP_016866060.1 | |||
| XM_017010572.1 | XP_016866061.1 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 | NM_001199917.1 | 14 | 2213 | NP_001186846.1 |
| NM_001199918.1 | 12 | 1875 | NP_001186847.1 | |
| NM_001199919.1 | NP_001186848.1 | |||
| NM_015599.2 | 13 | 2077 | NP_056414.1 | |
| XM_017010935.1 | XP_016866424.1 | |||
| XM_017010936.1 | XP_016866425.1 | |||
| XM_017010937.1 | XP_016866426.1 | |||
| XM_017010938.1 | XP_016866427.1 | |||
| AB032081.1 | BAB00613.1 | |||
| AF102265.1 | AAC72409.1 | |||
| AF180371.1 | AAD55097.1 | |||
| AK001365.1 | 12 | 1816 | ||
| AK023432.1 | ||||
| AK023709.1 | 3 | 703 | BAB14652.1 | |
| AK225363.1 | ||||
| AL117443.1 | 4 | 788 | CAB55928.1 | |
| BC001258.1 | AAH01258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map