Assay Details
Target Gene Details
Entrez Gene ID: | 929 |
Gene Name: | CD14 molecule |
Gene Aliases: |
- |
Location: |
Chr.5:140631728-140633701 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD14 | NM_000591.3 | 1 | 65 | NP_000582.1 |
| NM_001040021.2 | NP_001035110.1 | |||
| NM_001174104.1 | NP_001167575.1 | |||
| NM_001174105.1 | NP_001167576.1 | |||
| BP300412.1 | ||||
| CF593452.1 | 1 | 316 | ||
| DA269805.1 | ||||
| DA426297.1 | ||||
| DA681405.1 |
Target Gene Details
Entrez Gene ID: | 55374 |
Gene Name: | transmembrane and coiled-coil domains 6 |
Gene Aliases: |
PRO1580 |
Location: |
Chr.5:140596529-140647411 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCO6 | XM_011537665.2 | XP_011535967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3606953 | Chr.5:140632977 - 140633916 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map