Assay Details
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | NP_001012267.1 | ||
| NM_001286969.1 | NP_001273898.1 | |||
| NM_001286971.1 | NP_001273900.1 | |||
| AL833701.1 |
Target Gene Details
Entrez Gene ID: | 64768 |
Gene Name: | inositol-pentakisphosphate 2-kinase |
Gene Aliases: |
C9orf12, INSP5K2, IP5K, IPK1, bA476B13.1 |
Location: |
Chr.9:92613184-92670265 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPPK | NM_022755.5 | 13 | 2384 | NP_073592.1 |
| XM_017015041.1 | 14 | 2454 | XP_016870530.1 | |
| AF351200.1 | 1 | 606 | ||
| AF351201.1 | ||||
| AF351202.1 | 13 | 2190 | AAK69692.1 | |
| AK023225.1 | 13 | 2174 | BAB14476.1 | |
| AK024267.1 | 11 | 1962 | BAB14866.1 | |
| AK096392.1 | 2 | 1702 | ||
| AK123694.1 | 4 | 2030 | ||
| BC026154.2 | 13 | 2386 | AAH26154.1 |
More Information
Set Membership: |
|

Genomic Map