Assay Details
Target Gene Details
Entrez Gene ID: | 6493 |
Gene Name: | single-minded family bHLH transcription factor 2 |
Gene Aliases: |
HMC13F06, HMC29C01, SIM, bHLHe15 |
Location: |
Chr.21:36699117-36750219 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIM2 | NM_005069.4 | 1 | 638 | NP_005060.1 |
| NM_009586.3 | 1 | 638 | NP_033664.2 | |
| XM_017028442.1 | 1 | 654 | XP_016883931.1 | |
| BC110444.2 | 1 | 50 | AAI10445.1 | |
| U80456.1 | 1 | 116 | AAB62396.1 | |
| U80457.1 | 1 | 116 | AAB62397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953361 | Chr.21:36689801 - 36710300 on Build GRCh38 | Deletion |
|
| nsv828876 | Chr.21:36685480 - 36735889 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map