Assay Details
Target Gene Details
Entrez Gene ID: | 5927 |
Gene Name: | lysine demethylase 5A |
Gene Aliases: |
RBBP-2, RBBP2, RBP2 |
Location: |
Chr.12:280057-389455 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM5A | NM_001042603.2 | 28 | 8196 | NP_001036068.1 |
| AK057703.1 | 1 | 640 | ||
| AL133026.1 | 1 | 1128 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050912 | Chr.12:262244 - 292982 on Build GRCh38 | Gain |
|
| esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
|
| nsv1050781 | Chr.12:212187 - 674733 on Build GRCh38 | Loss |
|
| nsv570 | Chr.12:257625 - 286657 on Build GRCh38 | Deletion |
|
| esv34939 | Chr.12:262620 - 323985 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map