Assay Details
Target Gene Details
Entrez Gene ID: | 55450 |
Gene Name: | calcium/calmodulin dependent protein kinase II inhibitor 1 |
Gene Aliases: |
PRO1489 |
Location: |
Chr.1:20482391-20486235 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK2N1 | NM_018584.5 | NP_061054.2 | ||
| BC020630.1 | ||||
| BI820748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv821943 | Chr.1:20479518 - 20492154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map