Assay Details
Target Gene Details
Entrez Gene ID: | 4642 |
Gene Name: | myosin ID |
Gene Aliases: |
PPP1R108, myr4 |
Location: |
Chr.17:32492522-32877173 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO1D | NM_015194.2 | 22 | 5308 | NP_056009.1 |
| AB018270.2 | 22 | 5019 | ||
| AK026907.1 | 1 | 2027 | ||
| AK026920.1 | 19 | 4604 | ||
| BC030796.2 | 4 | 2480 | AAH30796.2 | |
| BC146763.1 | 22 | 5019 | ||
| BX537615.1 | 23 | 3319 | CAD97800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519839 | Chr.17:32448743 - 32502646 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map