Assay Details
Target Gene Details
Entrez Gene ID: | 83856 |
Gene Name: | fibronectin type III and SPRY domain containing 1 like |
Gene Aliases: |
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1 |
Location: |
Chr.9:105442183-105552433 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSD1L | NM_001145313.2 | NP_001138785.1 | ||
| NM_001287191.1 | NP_001274120.1 | |||
| NM_001287192.1 | NP_001274121.1 | |||
| NM_031919.4 | NP_114125.1 | |||
| XM_005252254.2 | XP_005252311.1 | |||
| XM_005252257.3 | XP_005252314.1 | |||
| XM_005252260.3 | XP_005252317.1 | |||
| XM_011519077.2 | XP_011517379.1 | |||
| XM_011519078.2 | XP_011517380.2 | |||
| XM_011519079.2 | XP_011517381.2 | |||
| XM_017015182.1 | XP_016870671.1 | |||
| XM_017015183.1 | XP_016870672.1 | |||
| XM_017015185.1 | XP_016870674.1 | |||
| XM_017015187.1 | XP_016870676.1 | |||
| AF088031.1 | ||||
| AF316830.1 | AAK26748.1 | |||
| AK296433.1 | ||||
| AK299350.1 | ||||
| AK299491.1 | ||||
| AK309817.1 | ||||
| BP219536.1 | ||||
| BP229804.1 | ||||
| BP365251.1 | ||||
| DN999402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037750 | Chr.9:105265411 - 105489452 on Build GRCh38 | Gain |
|
| nsv1053973 | Chr.9:105436896 - 105537905 on Build GRCh38 | Loss |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|
| nsv615115 | Chr.9:105442981 - 105535902 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map