Assay Details
Target Gene Details
Entrez Gene ID: | 5965 |
Gene Name: | RecQ like helicase |
Gene Aliases: |
RECQL1, RecQ1 |
Location: |
Chr.12:21468910-21501669 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RECQL | NM_002907.3 | NP_002898.2 | ||
| NM_032941.2 | NP_116559.1 | |||
| XM_005253461.3 | XP_005253518.1 | |||
| XM_005253462.4 | XP_005253519.1 | |||
| XM_005253463.3 | XP_005253520.1 | |||
| XM_005253464.3 | XP_005253521.1 | |||
| AK291627.1 | ||||
| AY157499.1 | AAO61939.1 | |||
| BC001052.2 | AAH01052.1 | |||
| BT007119.1 | AAP35783.1 | |||
| D37984.1 | BAA07200.1 | |||
| L36140.1 | AAA60261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map