Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAALADL2 | NM_207015.2 | 2 | 213 | NP_996898.2 |
| XM_006713560.2 | 4 | 296 | XP_006713623.1 | |
| XM_011512612.2 | 2 | 3550 | XP_011510914.1 | |
| XM_011512613.1 | 2 | 358 | XP_011510915.1 | |
| XM_011512615.2 | 2 | 3551 | XP_011510917.1 | |
| XM_011512617.2 | 2 | 3548 | XP_011510919.1 | |
| XM_017006070.1 | 6 | 738 | XP_016861559.1 | |
| XM_017006071.1 | 7 | 808 | XP_016861560.1 | |
| XM_017006072.1 | 8 | 942 | XP_016861561.1 | |
| XM_017006073.1 | 6 | 541 | XP_016861562.1 | |
| XM_017006074.1 | 8 | 2960 | XP_016861563.1 | |
| XM_017006075.1 | 5 | 1685 | XP_016861564.1 | |
| XM_017006076.1 | 5 | 478 | XP_016861565.1 | |
| XM_017006077.1 | 7 | 1889 | XP_016861566.1 | |
| XM_017006078.1 | 5 | 1706 | XP_016861567.1 | |
| XM_017006079.1 | 3 | 1182 | XP_016861568.1 | |
| XM_017006080.1 | 3 | 1553 | XP_016861569.1 | |
| XM_017006081.1 | 2 | 3550 | XP_016861570.1 | |
| XM_017006082.1 | 5 | 1692 | XP_016861571.1 | |
| AJ607396.1 | 2 | 254 | CAE54974.2 | |
| AK308416.1 | 2 | 229 | ||
| AL832144.1 | 2 | 213 | ||
| BC057243.1 | 2 | 216 | AAH57243.1 |
Target Gene Details
Entrez Gene ID: | 100862679 |
Gene Name: | NAALADL2 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.3:175079312-175115242 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAALADL2-AS3 | NR_046390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829799 | Chr.3:175000333 - 175179508 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map