Assay Details
Target Gene Details
Entrez Gene ID: | 2021 |
Gene Name: | endonuclease G |
Gene Aliases: |
- |
Location: |
Chr.9:128818402-128822676 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENDOG | NM_004435.2 | NP_004426.2 | ||
| XM_011518347.2 | XP_011516649.1 | |||
| AK130973.1 | ||||
| BC004922.2 | AAH04922.1 | |||
| BC016351.1 | AAH16351.1 | |||
| X79444.1 | CAA55963.1 |
Target Gene Details
Entrez Gene ID: | 51490 |
Gene Name: | SPOUT domain containing methyltransferase 1 |
Gene Aliases: |
C9orf114, CENP-32, HSPC109 |
Location: |
Chr.9:128819651-128833309 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPOUT1 | NM_016390.3 | 12 | 1303 | NP_057474.2 |
| XM_017014804.1 | 13 | 1570 | XP_016870293.1 | |
| AF161459.1 | AAF29074.1 | |||
| AK303563.1 | ||||
| AL110193.1 | 12 | 1303 | ||
| BC010579.1 | 12 | 1265 | AAH10579.1 | |
| BC021273.1 | 12 | 1276 | ||
| BC033677.1 | 12 | 1271 | AAH33677.1 | |
| BC039590.1 | 12 | 1288 | AAH39590.1 | |
| BC046133.1 | 12 | 1268 | AAH46133.1 | |
| BC063644.1 | 12 | 1271 | AAH63644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615412 | Chr.9:128754559 - 128824961 on Build GRCh38 | Loss |
|
| nsv521120 | Chr.9:128803275 - 128829871 on Build GRCh38 | Loss |
|
| nsv615411 | Chr.9:128727172 - 128829871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map