Assay Details
Target Gene Details
Entrez Gene ID: | 9913 |
Gene Name: | SPT7-like STAGA complex gamma subunit |
Gene Aliases: |
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H |
Location: |
Chr.2:27642568-27663840 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT7L | NM_001282729.1 | 6 | 2276 | NP_001269658.1 |
| NM_001282730.1 | 6 | 2484 | NP_001269659.1 | |
| NM_001282731.1 | 6 | 2326 | NP_001269660.1 | |
| NM_001282732.1 | 5 | 1866 | NP_001269661.1 | |
| NM_014860.2 | 6 | 2271 | NP_055675.1 | |
| XM_005264672.4 | 6 | 2473 | XP_005264729.1 | |
| AB018307.1 | 6 | 2013 | ||
| AF197954.1 | AAG28523.1 | |||
| AF224759.1 | AAG47636.1 | |||
| AK000329.1 | ||||
| AK304889.1 | ||||
| BC074000.1 | AAH74000.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115953697] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map