Assay Details
Target Gene Details
Entrez Gene ID: | 51059 |
Gene Name: | family with sequence similarity 135 member B |
Gene Aliases: |
C8ORFK32 |
Location: |
Chr.8:138130023-138498390 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM135B | NM_015912.3 | 20 | 6852 | NP_056996.2 |
| XM_011517056.2 | XP_011515358.1 | |||
| XM_011517058.2 | XP_011515360.1 | |||
| XM_011517059.1 | XP_011515361.1 | |||
| XM_011517060.2 | XP_011515362.1 | |||
| XM_011517061.2 | XP_011515363.1 | |||
| XM_011517062.1 | XP_011515364.1 | |||
| XM_011517063.2 | XP_011515365.1 | |||
| XM_011517064.2 | XP_011515366.1 | |||
| XM_011517065.1 | XP_011515367.1 | |||
| XM_011517068.2 | XP_011515370.1 | |||
| XM_011517069.2 | XP_011515371.1 | |||
| XM_011517072.2 | XP_011515374.1 | |||
| XM_011517073.2 | XP_011515375.1 | |||
| XM_011517074.1 | XP_011515376.1 | |||
| XM_017013470.1 | XP_016868959.1 | |||
| XM_017013471.1 | XP_016868960.1 | |||
| CR936831.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831477 | Chr.8:137996707 - 138148120 on Build GRCh38 | Loss |
|
| nsv1030981 | Chr.8:137782191 - 138241562 on Build GRCh38 | Gain |
|
| esv3576609 | Chr.8:137771394 - 138238230 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map