Assay Details
Target Gene Details
Entrez Gene ID: | 2098 |
Gene Name: | esterase D |
Gene Aliases: |
FGH |
Location: |
Chr.13:46771256-46797681 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESD | NM_001984.1 | 10 | 1001 | NP_001975.1 |
| XM_005266278.2 | 10 | 1065 | XP_005266335.1 | |
| XM_011534954.1 | 10 | 953 | XP_011533256.1 | |
| XM_017020414.1 | 10 | 2275 | XP_016875903.1 | |
| AF112219.1 | 10 | 1001 | AAC99788.1 | |
| AK098540.1 | 10 | 930 | ||
| BC001169.1 | 10 | 916 | AAH01169.1 | |
| BT007059.1 | AAP35708.1 | |||
| M13450.1 | 9 | 863 | AAA52408.1 |
More Information
Set Membership: |
|

Genomic Map