Assay Details
Target Gene Details
Entrez Gene ID: | 163126 |
Gene Name: | EP300 interacting inhibitor of differentiation 2 |
Gene Aliases: |
CRI2, EID-2 |
Location: |
Chr.19:39538806-39540198 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EID2 | NM_153232.3 | 1 | 646 | NP_694964.3 |
| AK128468.1 | 2 | 500 | BAC87454.1 | |
| AY251272.1 | 1 | 596 | AAP12559.1 | |
| BC030137.2 | 1 | 632 | AAH30137.3 | |
| BC099899.1 | 1 | 399 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579509 | Chr.19:39514000 - 39642798 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map