Assay Details
Target Gene Details
Entrez Gene ID: | 84196 |
Gene Name: | ubiquitin specific peptidase 48 |
Gene Aliases: |
RAP1GA1, USP31 |
Location: |
Chr.1:21678298-21783620 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34155 | Chr.1:21367084 - 21840123 on Build GRCh38 | Loss |
|
| nsv834391 | Chr.1:21523440 - 21723989 on Build GRCh38 | Loss |
|
| nsv1008548 | Chr.1:21694652 - 21741766 on Build GRCh38 | Loss |
|
| nsv5854 | Chr.1:21676656 - 21711616 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map