Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLM1 | NM_016548.3 | 8 | 1085 | NP_057632.2 |
| NM_177937.2 | 8 | 1077 | NP_808800.1 | |
| AF236056.1 | 8 | 1026 | AAF44663.1 | |
| AK027717.1 | 8 | 1048 | ||
| AK074188.1 | 1 | 79 | BAB85012.1 | |
| AK075542.1 | 8 | 1019 | BAC11685.1 | |
| AK075557.1 | 8 | 989 | ||
| AK092943.1 | 9 | 965 | ||
| AY358593.1 | 8 | 990 | AAQ88956.1 | |
| BC001740.1 | 8 | 1000 | AAH01740.1 | |
| CR457201.1 | 7 | 876 | CAG33482.1 | |
| EU794616.1 | 8 | 1044 |
More Information
Set Membership: |
|

Genomic Map