Assay Details
Target Gene Details
Entrez Gene ID: | 25966 |
Gene Name: | C2 calcium dependent domain containing 2 |
Gene Aliases: |
C21orf25, C21orf258, TMEM24L |
Location: |
Chr.21:41885110-41953957 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD2 | NM_015500.1 | 14 | 4252 | NP_056315.1 |
| NM_199050.2 | 13 | 3751 | NP_950251.1 | |
| XM_005261109.4 | 13 | 3792 | XP_005261166.1 | |
| XM_011529522.1 | 14 | 4352 | XP_011527824.1 | |
| XM_011529523.2 | 15 | 5018 | XP_011527825.1 | |
| XM_017028315.1 | 11 | 3488 | XP_016883804.1 | |
| AB047784.1 | 14 | 4252 | BAD74068.1 | |
| AK125120.1 | 5 | 2821 | ||
| AK128550.1 | 11 | 3591 | ||
| AL050173.1 | 3 | 2466 | CAB43307.1 | |
| AL833323.1 | 13 | 3751 | ||
| CR627455.1 | 6 | 4026 | CAH10536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587575 | Chr.21:41887220 - 41907962 on Build GRCh38 | Loss |
|
| nsv828881 | Chr.21:41887014 - 41887850 on Build GRCh38 | Gain |
|
| nsv587574 | Chr.21:41877778 - 41893574 on Build GRCh38 | Loss |
|
| dgv7850n54 | Chr.21:41870944 - 41887220 on Build GRCh38 | Loss |
|
| nsv587564 | Chr.21:41833660 - 41887220 on Build GRCh38 | Loss |
|
| dgv7849n54 | Chr.21:41863326 - 41898934 on Build GRCh38 | Loss |
|

Genomic Map