Assay Details
Target Gene Details
Entrez Gene ID: | 84795 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 2 |
Gene Aliases: |
C10orf33, FP3420 |
Location: |
Chr.10:98383565-98415221 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD2 | NM_032709.2 | 10 | 1079 | NP_116098.2 |
| XM_011540293.2 | 11 | 2501 | XP_011538595.1 | |
| XM_011540301.2 | 3 | 1915 | XP_011538603.1 | |
| XM_017016835.1 | 10 | 1040 | XP_016872324.1 | |
| XM_017016836.1 | 10 | 1040 | XP_016872325.1 | |
| XM_017016837.1 | 10 | 1040 | XP_016872326.1 | |
| XM_017016838.1 | 10 | 3275 | XP_016872327.1 | |
| XM_017016839.1 | 8 | 1132 | XP_016872328.1 | |
| XM_017016840.1 | 8 | 986 | XP_016872329.1 | |
| XM_017016841.1 | 7 | 1293 | XP_016872330.1 | |
| AK074429.1 | 10 | 1040 | ||
| AK075265.1 | 10 | 1017 | BAC11507.1 | |
| AK225637.1 | 10 | 1033 | ||
| BC006131.1 | 10 | 1042 | AAH06131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515723 | Chr.10:98388419 - 98435072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map