Assay Details
Target Gene Details
Entrez Gene ID: | 3579 |
Gene Name: | C-X-C motif chemokine receptor 2 |
Gene Aliases: |
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB |
Location: |
Chr.2:218125290-218137253 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR2 | NM_001168298.1 | 4 | 2560 | NP_001161770.1 |
| NM_001557.3 | 3 | 2768 | NP_001548.1 | |
| XM_005246530.3 | 4 | 2540 | XP_005246587.1 | |
| XM_017003990.1 | 4 | 2520 | XP_016859479.1 | |
| XM_017003991.1 | 2 | 2431 | XP_016859480.1 | |
| XM_017003992.1 | 2 | 2701 | XP_016859481.1 | |
| AJ710879.1 | 1 | 34 | ||
| BC037961.1 | 3 | 2722 | AAH37961.2 | |
| L19593.1 | 3 | 2758 | AAB59437.1 |
More Information
Set Membership: |
|

Genomic Map