Assay Details
Target Gene Details
Entrez Gene ID: | 79612 |
Gene Name: | N(alpha)-acetyltransferase 16, NatA auxiliary subunit |
Gene Aliases: |
NARG1L |
Location: |
Chr.13:41311205-41377031 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA16 | NM_001110798.1 | NP_001104268.1 | ||
| NM_018527.3 | NP_060997.2 | |||
| NM_024561.4 | NP_078837.3 | |||
| XM_006719866.2 | XP_006719929.1 | |||
| XM_011535227.2 | XP_011533529.1 | |||
| XM_011535228.1 | XP_011533530.1 | |||
| XM_017020745.1 | XP_016876234.1 | |||
| XM_017020746.1 | XP_016876235.1 | |||
| AK097224.1 | ||||
| AK291248.1 | ||||
| BC032318.1 | AAH32318.1 | |||
| BC064592.1 | AAH64592.1 | |||
| BC101719.1 | ||||
| BC101721.1 | ||||
| BX640665.1 | CAE45801.1 | |||
| DB078940.1 |
More Information
Set Membership: |
|

Genomic Map