Assay Details
Target Gene Details
Entrez Gene ID: | 9214 |
Gene Name: | Fc fragment of IgM receptor |
Gene Aliases: |
FAIM3, TOSO |
Location: |
Chr.1:206903282-206922033 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCMR | NM_001142473.1 | 7 | 2069 | NP_001135945.1 |
| NM_001193338.1 | 7 | 2131 | NP_001180267.1 | |
| NM_005449.4 | 8 | 2405 | NP_005440.1 | |
| XM_005273351.3 | 7 | 2346 | XP_005273408.1 | |
| CB158779.1 | 1 | 174 |
Target Gene Details
Entrez Gene ID: | 11009 |
Gene Name: | interleukin 24 |
Gene Aliases: |
C49A, FISP, IL10B, MDA7, MOB5, ST16 |
Location: |
Chr.1:206897404-206904139 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL24 | NM_001185156.1 | 7 | 1806 | NP_001172085.1 |
| NM_001185157.1 | 6 | 1647 | NP_001172086.1 | |
| NM_001185158.1 | 5 | 1448 | NP_001172087.1 | |
| NM_006850.3 | 7 | 1803 | NP_006841.1 | |
| AI521306.1 | 1 | 230 | ||
| AY641440.1 | 6 | 1606 | AAV52800.1 | |
| AY641441.1 | 6 | 1646 | AAV52801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010486 | Chr.1:206876486 - 207031685 on Build GRCh38 | Gain |
|
| nsv468049 | Chr.1:206894495 - 206963993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map