Assay Details
Target Gene Details
Entrez Gene ID: | 170506 |
Gene Name: | DEAH-box helicase 36 |
Gene Aliases: |
DDX36, G4R1, MLEL1, RHAU |
Location: |
Chr.3:154275668-154324497 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX36 | NM_001114397.1 | 25 | 3358 | NP_001107869.1 |
| NM_020865.2 | 25 | 3400 | NP_065916.2 | |
| AB040921.1 | 23 | 2852 | BAA96012.1 | |
| AF217190.1 | 25 | 3392 | AAG36783.1 | |
| BC036035.1 | 24 | 3277 | AAH36035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763308 | Chr.3:154186065 - 154347347 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map