Assay Details
Target Gene Details
Entrez Gene ID: | 10575 |
Gene Name: | chaperonin containing TCP1 subunit 4 |
Gene Aliases: |
CCT-DELTA, Cctd, SRB |
Location: |
Chr.2:61868127-61888671 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT4 | NM_001256721.1 | NP_001243650.1 | ||
| NM_006430.3 | NP_006421.2 | |||
| AF026291.1 | AAC96010.1 | |||
| AK129860.1 | ||||
| AK290538.1 | ||||
| AK294665.1 | ||||
| AK295330.1 | ||||
| AK303082.1 | ||||
| AK307856.1 | ||||
| AK308942.1 | ||||
| AK312586.1 | ||||
| AK315975.1 | ||||
| BC106933.1 | AAI06934.1 | |||
| BC106934.1 | AAI06935.1 | |||
| BM765256.1 | ||||
| U38846.1 | AAC50384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582123 | Chr.2:61719360 - 62324337 on Build GRCh38 | Gain |
|
| nsv458074 | Chr.2:61692252 - 62020316 on Build GRCh38 | Gain |
|
| nsv470467 | Chr.2:61786803 - 62020316 on Build GRCh38 | Gain |
|
| nsv582118 | Chr.2:61431550 - 62075525 on Build GRCh38 | Gain |
|
| esv34000 | Chr.2:61672382 - 62157212 on Build GRCh38 | Loss |
|
| nsv1009446 | Chr.2:61872503 - 61972765 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map