Assay Details
Target Gene Details
Entrez Gene ID: | 65056 |
Gene Name: | GC-rich promoter binding protein 1 |
Gene Aliases: |
GPBP, SSH6, VASCULIN |
Location: |
Chr.5:57173826-57264679 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPBP1 | NM_001127235.2 | 1 | 286 | NP_001120707.2 |
| NM_001203246.1 | 2 | 550 | NP_001190175.1 | |
| NM_022913.3 | 2 | 550 | NP_075064.1 | |
| XM_006714673.3 | 2 | 590 | XP_006714736.1 | |
| XM_006714674.3 | 2 | 590 | XP_006714737.1 | |
| XM_017009754.1 | XP_016865243.1 | |||
| XM_017009755.1 | XP_016865244.1 | |||
| XM_017009756.1 | XP_016865245.1 | |||
| XM_017009757.1 | 2 | 590 | XP_016865246.1 | |
| XM_017009758.1 | 2 | 590 | XP_016865247.1 | |
| XM_017009759.1 | 2 | 387 | XP_016865248.1 | |
| AK311625.1 | 2 | 550 | ||
| AL161991.1 | 2 | 449 | CAB82324.2 | |
| AY226828.1 | 2 | 381 | AAO34124.1 | |
| BC113004.2 | 1 | 299 | AAI13005.1 | |
| DB089516.1 | ||||
| DB271841.1 |
More Information
Set Membership: |
|

Genomic Map