Assay Details
Target Gene Details
Entrez Gene ID: | 6367 |
Gene Name: | C-C motif chemokine ligand 22 |
Gene Aliases: |
A-152E5.1, ABCD-1, DC/B-CK, MDC, SCYA22, STCP-1 |
Location: |
Chr.16:57357909-57366190 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL22 | NM_002990.4 | 3 | 744 | NP_002981.2 |
| XM_017023531.1 | 4 | 764 | XP_016879020.1 | |
| BC027952.1 | 3 | 721 | AAH27952.1 | |
| U83171.1 | 3 | 729 | AAB58360.1 | |
| U83239.1 | 3 | 724 | AAB53372.1 |
More Information
Set Membership: |
|

Genomic Map