Assay Details
Target Gene Details
Entrez Gene ID: | 257177 |
Gene Name: | cilia and flagella associated protein 126 |
Gene Aliases: |
C1orf192, Flattop, Fltp |
Location: |
Chr.1:161364731-161367883 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP126 | NM_001013625.3 | NP_001013647.2 | ||
| DT215043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010776 | Chr.1:161312594 - 161439428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map