Assay Details
Target Gene Details
Entrez Gene ID: | 246175 |
Gene Name: | CCR4-NOT transcription complex subunit 6 like |
Gene Aliases: |
CCR4b |
Location: |
Chr.4:77713387-77820238 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT6L | NM_001286790.1 | 12 | 4307 | NP_001273719.1 |
| NM_144571.2 | 12 | 4236 | NP_653172.2 | |
| XM_011531806.2 | 13 | 4371 | XP_011530108.1 | |
| XM_011531807.2 | 13 | 4345 | XP_011530109.1 | |
| XM_011531808.2 | 12 | 4317 | XP_011530110.1 | |
| XM_011531809.1 | 12 | 4262 | XP_011530111.1 | |
| XM_011531810.1 | 12 | 4822 | XP_011530112.1 | |
| XM_017007956.1 | 12 | 4259 | XP_016863445.1 | |
| XM_017007957.1 | 12 | 4585 | XP_016863446.1 | |
| AL133112.1 | 6 | 3534 | CAB61415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518757 | Chr.4:77716111 - 77773984 on Build GRCh38 | Loss |
|
| nsv829976 | Chr.4:77655503 - 77836644 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map