Assay Details
Target Gene Details
Entrez Gene ID: | 3672 |
Gene Name: | integrin subunit alpha 1 |
Gene Aliases: |
CD49a, VLA1 |
Location: |
Chr.5:52788302-52953655 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA1 | NM_181501.1 | NP_852478.1 | ||
| AK300423.1 | ||||
| BC137121.1 | ||||
| BC137122.1 |
Target Gene Details
Entrez Gene ID: | 53918 |
Gene Name: | pelota homolog (Drosophila) |
Gene Aliases: |
CGI-17, PRO1770 |
Location: |
Chr.5:52787940-52802617 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PELO | NM_015946.4 | 3 | 2434 | NP_057030.3 |
| AK025729.1 | 2 | 1748 | ||
| AY117399.1 | 3 | 2434 | AAM89414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830295 | Chr.5:52788698 - 52961866 on Build GRCh38 | Gain |
|
| esv3894188 | Chr.5:52769347 - 52857921 on Build GRCh38 | Gain |
|
| nsv471013 | Chr.5:52483363 - 53043284 on Build GRCh38 | Gain |
|
| nsv1020640 | Chr.5:52762996 - 52825024 on Build GRCh38 | Gain |
|
| nsv1027410 | Chr.5:52726879 - 52807526 on Build GRCh38 | Gain |
|
| nsv462187 | Chr.5:52462970 - 53037590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map