Assay Details
Target Gene Details
Entrez Gene ID: | 25978 |
Gene Name: | charged multivesicular body protein 2B |
Gene Aliases: |
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B |
Location: |
Chr.3:87227263-87255548 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP2B | NM_001244644.1 | 2 | 437 | NP_001231573.1 |
| NM_014043.3 | 3 | 529 | NP_054762.2 | |
| XM_011533576.2 | 3 | 360 | XP_011531878.1 | |
| AF151842.1 | 3 | 488 | AAD34079.1 | |
| AK002165.1 | 3 | 502 | ||
| AK002180.1 | 3 | 507 | ||
| AK222654.1 | 3 | 489 | BAD96374.1 | |
| AK296072.1 | 2 | 429 | ||
| AK316585.1 | 3 | 493 | ||
| AL080122.1 | 3 | 502 | CAB45721.1 | |
| BC001553.1 | 3 | 345 | AAH01553.1 | |
| BI821848.1 | 3 | 506 | ||
| BM828410.1 | 3 | 529 | ||
| CR533456.1 | 3 | 269 | CAG38487.1 | |
| DA426159.1 | 3 | 500 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953231 | Chr.3:87227951 - 87252550 on Build GRCh38 | Duplication |
|
| nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain |
|
| esv3568885 | Chr.3:87182571 - 87518398 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map