Assay Details
Target Gene Details
Entrez Gene ID: | 50515 |
Gene Name: | carbohydrate (chondroitin 4) sulfotransferase 11 |
Gene Aliases: |
C4ST, C4ST-1, C4ST1, HSA269537 |
Location: |
Chr.12:104456914-104762014 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHST11 | NM_001173982.1 | 1 | 323 | NP_001167453.1 |
| NM_018413.5 | 1 | 323 | NP_060883.1 | |
| AB042326.1 | 1 | 211 | BAA95485.1 | |
| AF239820.1 | AAF81691.1 | |||
| AJ289134.1 | CAB92134.1 | |||
| AK290923.1 | 1 | 264 | ||
| AK313217.1 | 1 | 266 | ||
| BC013315.1 | AAH13315.1 | |||
| DA853534.1 | 1 | 323 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832502 | Chr.12:104439951 - 104514510 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map