Assay Details
Target Gene Details
Entrez Gene ID: | 56676 |
Gene Name: | achaete-scute family bHLH transcription factor 3 |
Gene Aliases: |
HASH3, SGN1, bHLHa42 |
Location: |
Chr.11:8937572-8943033 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL3 | NM_020646.2 | NP_065697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625324 | Chr.11:8936425 - 8943806 on Build GRCh38 | Loss |
|
| nsv520248 | Chr.11:8942772 - 8943391 on Build GRCh38 | Loss |
|
| dgv191e199 | Chr.11:8936445 - 8943662 on Build GRCh38 | Deletion |
|
| nsv435923 | Chr.11:8817778 - 8979055 on Build GRCh38 | Deletion |
|
| dgv1640n54 | Chr.11:8937356 - 8943391 on Build GRCh38 | Loss |
|
| esv2759803 | Chr.11:8763266 - 8988026 on Build GRCh38 | Gain |
|
| nsv832062 | Chr.11:8827748 - 8987925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map