Assay Details
Target Gene Details
Entrez Gene ID: | 63899 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 3 |
Gene Aliases: |
MST077, MSTP077 |
Location: |
Chr.3:94063011-94126786 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN3 | NM_022072.3 | 3 | 506 | NP_071355.1 |
| AF169972.1 | 3 | 514 | AAQ13600.1 | |
| AK026262.1 | 3 | 498 | BAB15417.1 | |
| BC020602.1 | 3 | 506 | AAH20602.1 | |
| BC057238.1 | 3 | 510 | AAH57238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss |
|
| esv3597014 | Chr.3:94083460 - 94086595 on Build GRCh38 | Loss |
|
| esv2678302 | Chr.3:94083472 - 94086613 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map