Assay Details
Target Gene Details
Entrez Gene ID: | 10210 |
Gene Name: | TOP1 binding arginine/serine rich protein |
Gene Aliases: |
LUN, P53BP3, RP31, TP53BPL |
Location: |
Chr.9:32540544-32552628 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPORS | NM_001195622.1 | 2 | 3348 | NP_001182551.1 |
| NM_005802.4 | 3 | 3543 | NP_005793.2 | |
| AB045732.1 | 3 | 3470 | BAB03714.1 | |
| AB045733.1 | 2 | 3275 | BAB03715.1 | |
| AI934577.1 | ||||
| AV712580.1 | 1 | 62 | ||
| BC060884.1 | 3 | 3483 | AAH60884.1 | |
| BQ007035.1 | 1 | 646 | ||
| BU674865.1 | 1 | 379 | ||
| U82939.1 | 2 | 3201 | AAC98530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map