Assay Details
Target Gene Details
Entrez Gene ID: | 10672 |
Gene Name: | G protein subunit alpha 13 |
Gene Aliases: |
G13 |
Location: |
Chr.17:65009289-65056802 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNA13 | NM_001282425.1 | 4 | 1225 | NP_001269354.1 |
| NM_006572.5 | 4 | 1535 | NP_006563.2 | |
| AK002117.1 | ||||
| AK302400.1 | 4 | 1225 | ||
| BC036756.1 | 4 | 1380 | AAH36756.1 | |
| L22075.1 | 4 | 1367 | AAA74235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066765 | Chr.17:64520760 - 65128185 on Build GRCh38 | Gain |
|
| nsv952361 | Chr.17:65009183 - 65052982 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map