Assay Details
Target Gene Details
Entrez Gene ID: | 262 |
Gene Name: | adenosylmethionine decarboxylase 1 |
Gene Aliases: |
ADOMETDC, AMD, SAMDC |
Location: |
Chr.6:110814621-110895713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMD1 | NM_001287215.1 | NP_001274144.1 | ||
| AK304351.1 |
Target Gene Details
Entrez Gene ID: | 23097 |
Gene Name: | cyclin dependent kinase 19 |
Gene Aliases: |
CDC2L6, CDK11, bA346C16.3 |
Location: |
Chr.6:110609978-110816531 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK19 | NM_001300960.1 | NP_001287889.1 | ||
| NM_001300963.1 | NP_001287892.1 | |||
| NM_001300964.1 | NP_001287893.1 | |||
| NM_015076.4 | NP_055891.1 | |||
| XM_011535630.2 | XP_011533932.1 | |||
| XM_011535631.2 | XP_011533933.1 | |||
| XM_017010587.1 | XP_016866076.1 | |||
| XM_017010588.1 | XP_016866077.1 | |||
| AB028951.2 | BAA82980.2 | |||
| AJ420571.1 | ||||
| AK292360.1 | ||||
| AY028424.1 | AAK27731.1 | |||
| BC024247.1 | AAH24247.1 | |||
| BC037289.1 | AAH37289.1 | |||
| DB030002.1 | ||||
| JX028238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain |
|
| nsv830773 | Chr.6:110813116 - 110985229 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map