Assay Details
Target Gene Details
Entrez Gene ID: | 57763 |
Gene Name: | ankyrin repeat family A member 2 |
Gene Aliases: |
ANKRA |
Location: |
Chr.5:73552200-73565705 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRA2 | NM_023039.4 | 1 | 262 | NP_075526.1 |
| XM_005248560.2 | 1 | 281 | XP_005248617.1 | |
| XM_017009678.1 | 1 | 281 | XP_016865167.1 | |
| AF251051.1 | 1 | 250 | AAK34941.1 | |
| AK022876.1 | BAB14288.1 | |||
| AL558295.3 | 1 | 262 | ||
| BC012917.1 | 1 | 128 | AAH12917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830346 | Chr.5:73503223 - 73642429 on Build GRCh38 | Loss |
|
| nsv830347 | Chr.5:73517327 - 73706624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map