Assay Details
Target Gene Details
Entrez Gene ID: | 8398 |
Gene Name: | phospholipase A2 group VI |
Gene Aliases: |
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta |
Location: |
Chr.22:38111495-38192099 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G6 | NM_001004426.1 | 12 | 1787 | NP_001004426.1 |
| NM_001199562.1 | 12 | 1742 | NP_001186491.1 | |
| NM_003560.2 | 13 | 1949 | NP_003551.2 | |
| XM_005261764.2 | 13 | 1924 | XP_005261821.1 | |
| XM_005261765.1 | 13 | 1921 | XP_005261822.1 | |
| XM_005261766.1 | 13 | 2016 | XP_005261823.1 | |
| XM_006724332.3 | 13 | 1925 | XP_006724395.1 | |
| XM_011530422.1 | 13 | 1800 | XP_011528724.1 | |
| XM_011530423.1 | 14 | 2036 | XP_011528725.1 | |
| XM_011530424.1 | 15 | 2160 | XP_011528726.1 | |
| XM_011530425.1 | 13 | 1860 | XP_011528727.1 | |
| XM_017028981.1 | 12 | 1854 | XP_016884470.1 | |
| XM_017028982.1 | 12 | 1759 | XP_016884471.1 | |
| XM_017028983.1 | 14 | 1998 | XP_016884472.1 | |
| XM_017028984.1 | 13 | 1874 | XP_016884473.1 | |
| XM_017028985.1 | 12 | 1692 | XP_016884474.1 | |
| AF064594.1 | 13 | 1949 | AAC97486.1 | |
| AF102988.1 | 11 | 1651 | AAD41722.1 | |
| AF102989.1 | 12 | 1813 | AAD41723.1 | |
| AK096521.1 | 12 | 1482 | ||
| AK128862.1 | 3 | 2312 | ||
| AK290825.1 | 12 | 1744 | ||
| AK291212.1 | 13 | 1935 | ||
| AK308308.1 | 12 | 1755 | ||
| AL080187.1 | 13 | 1949 | CAB45768.2 | |
| BC036742.1 | 13 | 1943 | AAH36742.2 | |
| BC051904.1 | 12 | 1723 | AAH51904.1 | |
| CR456543.1 | 13 | 1877 | CAG30429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss |
|
| nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss |
|
| nsv508732 | Chr.22:38096355 - 38180191 on Build GRCh38 | Insertion |
|
| nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map