Assay Details
Target Gene Details
Entrez Gene ID: | 83452 |
Gene Name: | RAB33B, member RAS oncogene family |
Gene Aliases: |
SMC2 |
Location: |
Chr.4:139453232-139475916 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB33B | NM_031296.2 | 2 | 1411 | NP_112586.1 |
| XM_011532299.1 | 3 | 1507 | XP_011530601.1 | |
| AF350420.1 | 2 | 1133 | AAL83916.1 | |
| AL136904.1 | 2 | 1411 | CAB66838.1 | |
| BC036064.1 | 2 | 1043 | AAH36064.1 | |
| BC111977.1 | 2 | 1063 | AAI11978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830084 | Chr.4:139326103 - 139478732 on Build GRCh38 | Loss |
|
| nsv830085 | Chr.4:139393507 - 139586168 on Build GRCh38 | Loss |
|
| nsv1023892 | Chr.4:139377986 - 139483575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map