Assay Details
Target Gene Details
Entrez Gene ID: | 51026 |
Gene Name: | golgi transport 1B |
Gene Aliases: |
CGI-141, GCT2, GOT1, GOT1B, YMR292W |
Location: |
Chr.12:21501765-21518403 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLT1B | NM_016072.4 | 3 | 346 | NP_057156.1 |
| XM_017019384.1 | 3 | 330 | XP_016874873.1 | |
| AB097020.1 | 3 | 296 | BAC77373.1 | |
| AF068292.1 | 3 | 321 | AAF65181.1 | |
| AF151899.1 | 3 | 280 | AAD34136.1 | |
| AK309736.1 | 2 | 226 | ||
| AK310889.1 | 4 | 448 | ||
| AK311331.1 | ||||
| AK311920.1 | 3 | 258 | ||
| AL136571.1 | 3 | 245 | CAB66506.1 | |
| AY358975.1 | 3 | 263 | AAQ89334.1 | |
| BC012455.1 | 3 | 267 | AAH12455.1 | |
| CR457389.1 | 3 | 187 | CAG33670.1 | |
| DA579356.1 | 3 | 346 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map