Assay Details
Target Gene Details
Entrez Gene ID: | 51564 |
Gene Name: | histone deacetylase 7 |
Gene Aliases: |
HD7, HD7A, HDAC7A |
Location: |
Chr.12:47782710-47820612 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC7 | NM_001098416.3 | 25 | 3583 | NP_001091886.1 |
| NM_001308090.1 | 25 | 3643 | NP_001295019.1 | |
| NM_015401.4 | 26 | 3694 | NP_056216.2 | |
| XM_011538478.1 | 26 | 3646 | XP_011536780.1 | |
| XM_011538479.1 | 27 | 4088 | XP_011536781.1 | |
| XM_011538480.1 | 25 | 3535 | XP_011536782.1 | |
| XM_011538481.1 | 26 | 3539 | XP_011536783.1 | |
| XM_011538482.1 | 26 | 3623 | XP_011536784.1 | |
| XM_017019455.1 | 25 | 3595 | XP_016874944.1 | |
| AK001032.1 | 10 | 1434 | BAA91474.1 | |
| AK001190.1 | 18 | 2616 | BAA91545.1 | |
| AK024469.1 | 21 | 3674 | BAB15759.1 | |
| AK026767.1 | 2 | 1449 | BAB15546.1 | |
| AK027781.1 | 8 | 1333 | BAB55363.1 | |
| AK122588.1 | 11 | 5822 | BAC56929.1 | |
| AK128383.1 | 18 | 2537 | ||
| AK225910.1 | ||||
| AL117455.1 | 22 | 3161 | CAB55935.1 | |
| AY321367.1 | 25 | 3468 | AAP84704.1 | |
| BC006453.1 | 15 | 2118 | ||
| BC020505.2 | 18 | 2661 | AAH20505.2 | |
| BC064840.1 | 25 | 3477 | AAH64840.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470289 | Chr.12:47716768 - 47786725 on Build GRCh38 | Gain |
|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map